siRNA Details
| Gene Symbol: Helz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198298.1 | NP_938040.1 | 14 | 1876 | ||
| GenBank mRNA | AB093209.1 | - | 15 | 1937 | ||
| BC076626.1 | - | 14 | 1876 | 30640711 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Helz| Gene Symbol: Helz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198298.1 | NP_938040.1 | 14 | 1876 | ||
| GenBank mRNA | AB093209.1 | - | 15 | 1937 | ||
| BC076626.1 | - | 14 | 1876 | 30640711 | ||