siRNA Details
| Gene Symbol: Skap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033186.3 | NP_001028358.1 | 5 | 428 | ||
| NM_001177898.1 | NP_001171369.1 | 5 | 428 | |||
| NM_001177899.1 | NP_001171370.1 | 5 | 428 | |||
| GenBank mRNA | AK040068.1 | - | 5 | 428 | ||
| AK137942.1 | - | 5 | 396 | |||
| AK138030.1 | - | 5 | 411 | |||
| AK169825.1 | - | 5 | 393 | |||
| BC145402.1 | - | 5 | 357 | 9053960 | ||
| BC150764.1 | - | 5 | 357 | 9087675 | ||
| BC171961.1 | - | 5 | 357 | 9054247 | ||
| BY723539.1 | - | 5 | 408 | |||

Skap1
Genomic Map