siRNA Details
| Gene Symbol: 1700061G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030141.1 | NP_084417.1 | 10 | 1499 | ||
| GenBank mRNA | AK028078.1 | - | 10 | 1650 | ||
| BC116283.1 | - | 10 | 1499 | 40094427 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700061G19Rik| Gene Symbol: 1700061G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030141.1 | NP_084417.1 | 10 | 1499 | ||
| GenBank mRNA | AK028078.1 | - | 10 | 1650 | ||
| BC116283.1 | - | 10 | 1499 | 40094427 | ||