siRNA Details
| Gene Symbol: Naa35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030153.2 | NP_084429.2 | 5 | 436 | ||
| GenBank mRNA | AK021042.1 | - | 5 | 327 | ||
| AK210077.1 | - | 2 | 89 | |||
| AY102701.1 | - | 5 | 358 | |||
| BC027201.1 | - | 5 | 372 | |||
| BC043701.1 | - | 5 | 349 | |||
| BC056435.1 | - | 5 | 345 | 5707801 | ||

Naa35
Genomic Map