siRNA Details
| Gene Symbol: Naa35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030153.2 | NP_084429.2 | 7 | 667 | ||
| GenBank mRNA | AK021042.1 | - | 7 | 558 | ||
| AY102701.1 | - | 7 | 589 | |||
| BC027201.1 | - | 7 | 603 | |||
| BC043701.1 | - | 7 | 580 | |||
| BC056435.1 | - | 7 | 576 | 5707801 | ||

Naa35
Genomic Map