siRNA Details
| Gene Symbol: Naa35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030153.2 | NP_084429.2 | 15 | 1390 | ||
| GenBank mRNA | AK050805.1 | - | 5 | 707 | ||
| AK177551.1 | - | 2 | 51 | |||
| AY102701.1 | - | 15 | 1312 | |||
| BC056435.1 | - | 15 | 1299 | 5707801 | ||

Naa35
Genomic Map