siRNA Details
| Gene Symbol: Filip1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040397.4 | NP_001035487.2 | 5 | 2494 | ||
| NM_001177871.1 | NP_001171342.1 | 2 | 1801 | |||
| GenBank mRNA | AK019472.2 | - | 2 | 1802 | ||
| AK132319.1 | - | 2 | 1802 | |||
| BC062166.1 | - | 2 | 1784 | |||
| BC098507.1 | - | 1 | 1638 | |||
| EU623548.1 | - | 1 | 1613 | |||
| EU704258.1 | - | 5 | 2341 | |||

Filip1l
Genomic Map