siRNA Details
| Gene Symbol: Zc3h6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178404.3 | NP_848491.2 | 12 | 2680 | ||
| GenBank mRNA | AK019473.1 | - | 5 | 1225 | ||
| AK039171.1 | - | 12 | 2675 | |||
| AK220224.1 | - | 3 | 616 | |||
| BC043311.1 | - | 8 | 1636 | 5067309 | ||

Zc3h6
Genomic Map