siRNA Details
| Gene Symbol: Galnt15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030166.3 | NP_084442.1 | 3 | 1284 | ||
| GenBank mRNA | AB175682.1 | - | 3 | 894 | ||
| AK019470.1 | - | 3 | 1399 | |||
| AK030347.1 | - | 3 | 537 | |||
| AK163883.1 | - | 3 | 1160 | |||
| BC133711.1 | - | 3 | 1284 | 40143107 | ||

Galnt15
Genomic Map