siRNA Details
| Gene Symbol: Mctp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030174.2 | NP_084450.2 | 10 | 1283 | ||
| GenBank mRNA | AK047562.1 | - | 10 | 1284 | ||
| AK054478.1 | - | 6 | 656 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mctp1| Gene Symbol: Mctp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030174.2 | NP_084450.2 | 10 | 1283 | ||
| GenBank mRNA | AK047562.1 | - | 10 | 1284 | ||
| AK054478.1 | - | 6 | 656 | |||