siRNA Details
| Gene Symbol: Brpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282126.1 | NP_001269055.1 | 2 | 768 | ||
| NM_001282127.1 | NP_001269056.1 | 2 | 536 | |||
| NM_001282128.1 | NP_001269057.1 | 2 | 768 | |||
| NM_030178.2 | NP_084454.1 | 2 | 536 | |||
| GenBank mRNA | AK043372.1 | - | 2 | 769 | ||
| AK156375.1 | - | 2 | 716 | |||
| AK206283.1 | - | 1 | 245 | |||
| BC046521.1 | - | 2 | 476 | 5363697 | ||
| BC138361.1 | - | 2 | 360 | 8861381 | ||
| BC145256.1 | - | 2 | 592 | 9053809 | ||
| DQ288860.1 | - | 1 | 325 | |||

Brpf1
Genomic Map