siRNA Details
| Gene Symbol: Ndor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082476.2 | NP_001075945.1 | 2 | 232 | ||
| NM_001252541.1 | NP_001239470.1 | 2 | 232 | |||
| NM_001252542.1 | NP_001239471.1 | 2 | 299 | |||
| GenBank mRNA | AK019617.1 | - | 2 | 229 | ||
| AK143124.1 | - | 2 | 272 | |||
| AK163207.1 | - | 2 | 202 | |||
| AK169885.1 | - | 2 | 291 | |||
| AK211498.1 | - | 2 | 150 | |||
| BC049789.1 | - | 2 | 282 | 6331033 | ||
| BQ771207.1 | - | 2 | 199 | |||
| CD354207.1 | - | 623 | ||||

Ndor1
Genomic Map