siRNA Details
| Gene Symbol: Slc25a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172436.3 | NP_766024.1 | 16 | 1716 | ||
| GenBank mRNA | AK019150.1 | - | 16 | 1719 | ||
| AK030595.1 | - | 16 | 1746 | |||
| AK083156.1 | - | 16 | 1717 | |||
| AK144958.1 | - | 16 | 1751 | |||
| AK147948.1 | - | 16 | 1732 | |||
| BC013845.1 | - | 16 | 1695 | |||
| BC024952.1 | - | 16 | 1909 | |||
| BC060505.1 | - | 16 | 1703 | 6311836 | ||

Slc25a12
Genomic Map