siRNA Details
| Gene Symbol: Scyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023912.2 | NP_076401.1 | 8 | 1225 | ||
| GenBank mRNA | AF276514.1 | - | 8 | 1225 | ||
| AK019240.1 | - | 8 | 1218 | |||
| AK134356.1 | - | 8 | 1224 | |||
| AK144134.1 | - | 8 | 1211 | |||
| AK145869.1 | - | 8 | 1245 | |||
| AK161813.1 | - | 8 | 1224 | |||
| BC034519.1 | - | 8 | 1211 | 3672524 | ||
| HM856325.1 | - | 8 | 1225 | |||
| HM856326.1 | - | 8 | 1225 | |||

Scyl1
Genomic Map