siRNA Details
| Gene Symbol: Misp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030218.2 | NP_084494.1 | 2 | 1277 | ||
| GenBank mRNA | AK020266.1 | - | 2 | 1277 | ||
| AK146817.1 | - | 2 | 1298 | |||
| AK147132.1 | - | 2 | 1265 | |||
| AK168406.1 | - | 2 | 1304 | |||
| BC013508.1 | - | 2 | 1255 | 4225010 | ||

Misp
Genomic Map