siRNA Details
| Gene Symbol: Igsf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207205.2 | NP_997088.2 | 6, 7 | 2334 | ||
| GenBank mRNA | AK045721.1 | - | 3, 4 | 1064 | ||
| AK084011.1 | - | 6, 7 | 2372 | |||
| AK129151.1 | - | 6, 7 | 2334 | |||
| BC052892.1 | - | 5, 6 | 1758 | 30055632 | ||

Igsf3
Genomic Map