siRNA Details
| Gene Symbol: Asb15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080847.3 | NP_543123.2 | 4 | 372 | ||
| GenBank mRNA | AF403043.1 | - | 4 | 373 | ||
| AK019556.1 | - | 4 | 373 | |||
| AK029759.1 | - | 4 | 304 | |||
| AK079418.1 | - | 4 | 478 | |||
| BC050794.1 | - | 4 | 321 | 6742480 | ||

Asb15
Genomic Map