siRNA Details
| Gene Symbol: Sp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080964.1 | NP_001074433.1 | 3 | 782 | ||
| NM_030220.3 | NP_084496.2 | 3 | 785 | |||
| GenBank mRNA | AK019649.2 | - | 4 | 998 | ||
| AK078246.1 | - | 3 | 765 | |||
| AK080877.1 | - | 3 | 793 | |||
| AK139302.1 | - | 3 | 752 | |||
| AK156580.1 | - | 3 | 767 | |||
| AK172877.1 | - | 3 | 756 | |||
| BC021759.1 | - | 3 | 752 | 5151210 | ||
| BC086457.1 | - | 3 | 751 | 6490946 | ||
| GU126673.1 | - | 3 | 741 | |||

Sp2
Genomic Map