siRNA Details
| Gene Symbol: Sp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080964.1 | NP_001074433.1 | 7 | 1812 | ||
| NM_030220.3 | NP_084496.2 | 7 | 1815 | |||
| GenBank mRNA | AK019649.2 | - | 8 | 2028 | ||
| AK078246.1 | - | 7 | 1795 | |||
| AK080877.1 | - | 7 | 1823 | |||
| AK139302.1 | - | 7 | 1782 | |||
| AK156580.1 | - | 7 | 1797 | |||
| AK172877.1 | - | 7 | 1786 | |||
| BC021759.1 | - | 7 | 1782 | 5151210 | ||
| BC086457.1 | - | 7 | 1781 | 6490946 | ||
| GU126673.1 | - | 7 | 1771 | |||

Sp2
Genomic Map