siRNA Details
| Gene Symbol: Ltn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081068.1 | NP_001074537.1 | 2 | 237 | ||
| GenBank mRNA | AK019706.2 | - | 2 | 168 | ||
| AK036725.1 | - | 2 | 167 | |||
| AK165062.1 | - | 2 | 158 | |||
| AK173009.1 | - | 2 | 108 | |||
| BC116272.2 | - | 2 | 191 | |||
| BC116273.2 | - | 2 | 194 | |||

Ltn1
Genomic Map