siRNA Details
Gene Symbol: Gas2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190406.1 | NP_001177335.1 | 3 | 853 | ||
NM_001190408.1 | NP_001177337.1 | 3 | 853 | |||
NM_030228.3 | NP_084504.2 | 4 | 1111 | |||
NM_144560.3 | NP_653146.1 | 4 | 1111 | |||
GenBank mRNA | AF508323.1 | - | 2 | 755 | ||
AF508324.1 | - | 2 | 755 | |||
AK019661.1 | - | 4 | 1086 | |||
AK052229.1 | - | 4 | 1112 | |||
AK082864.1 | - | 3 | 821 | |||
AK180368.1 | - | 2 | 126 | |||
AK218035.1 | - | 2 | 226 | |||
BC031785.1 | - | 3 | 803 | 4194333 |
