siRNA Details
Gene Symbol: Fbxo34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030236.3 | NP_084512.2 | 1, 2 | 155 | ||
GenBank mRNA | AK020018.1 | - | 1, 2 | 156 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fbxo34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030236.3 | NP_084512.2 | 1, 2 | 155 | ||
GenBank mRNA | AK020018.1 | - | 1, 2 | 156 |