siRNA Details
| Gene Symbol: Fbxo34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146085.1 | NP_001139557.1 | 4 | 1259 | ||
| NM_001146086.1 | NP_001139558.1 | 3 | 1182 | |||
| NM_030236.3 | NP_084512.2 | 2 | 1072 | |||
| GenBank mRNA | AK020018.1 | - | 2 | 1073 | ||
| AK077162.1 | - | 1 | 225 | |||
| AK171630.1 | - | 7 | 1607 | |||
| BC048149.1 | - | 4 | 1293 | 5706344 | ||
| BC049155.1 | - | 3 | 1124 | |||

Fbxo34
Genomic Map