siRNA Details
Gene Symbol: Trim39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024468.2 | NP_077788.2 | 6 | 1003 | ||
NM_178281.1 | NP_840065.1 | 6 | 988 | |||
GenBank mRNA | AB046382.1 | - | 6 | 1003 | ||
AK145861.1 | - | 6 | 939 | |||
AK166148.1 | - | 6 | 868 | |||
AK220298.1 | - | 6 | 924 | |||
BC031540.1 | - | 5 | 1045 | 3666785 |
