siRNA Details
| Gene Symbol: Bhlhe41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271768.1 | NP_001258697.1 | 3 | 293 | ||
| NM_024469.2 | NP_077789.1 | 3 | 293 | |||
| GenBank mRNA | AA013582.1 | - | 3 | 294 | ||
| AB044090.1 | - | 3 | 213 | |||
| AK170567.1 | - | 1 | 823 | |||
| BC023231.1 | - | 3 | 245 | |||
| BC037742.1 | - | 3 | 253 | |||
| BC051555.1 | - | 3 | 254 | |||
| BE985179.1 | - | 3 | 442 | |||

Bhlhe41
Genomic Map