siRNA Details
| Gene Symbol: Bhlhe41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271768.1 | NP_001258697.1 | 1 | 195 | ||
| NM_024469.2 | NP_077789.1 | 1 | 195 | |||
| GenBank mRNA | AA013582.1 | - | 1 | 196 | ||
| AB044090.1 | - | 1 | 115 | |||
| AK080341.1 | - | 1 | 128 | |||
| BC023231.1 | - | 1 | 147 | |||
| BC037742.1 | - | 1 | 155 | |||
| BC051555.1 | - | 1 | 156 | |||
| BE985179.1 | - | 5 | 540 | |||

Bhlhe41
Genomic Map