siRNA Details
| Gene Symbol: BC005537 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024473.3 | NP_077793.1 | 5 | 2400 | ||
| GenBank mRNA | AK035718.1 | - | 5 | 2400 | ||
| AK050530.1 | - | 1195 | ||||
| AK078765.1 | - | 5 | 2066 | |||
| AK160094.1 | - | 1357 | ||||
| BC002217.1 | - | 774 | 3490515 | |||
| BC005537.1 | - | 2037 | ||||

BC005537
Genomic Map