siRNA Details
| Gene Symbol: Parp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030253.3 | NP_084529.1 | 8 | 2389 | ||
| GenBank mRNA | AK037903.1 | - | 8 | 1956 | ||
| AK050032.1 | - | 8 | 1833 | |||
| BC003281.1 | - | 8 | 1831 | 3581841 | ||
| BC010312.1 | - | 7 | 1814 | |||
| BC070466.1 | - | 8 | 2389 | 30615225 | ||

Parp9
Genomic Map