siRNA Details
| Gene Symbol: Lysmd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030257.1 | NP_084533.1 | 3 | 878 | ||
| GenBank mRNA | AK029514.1 | - | 3 | 797 | ||
| BC003322.1 | - | 3 | 878 | 3156298 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lysmd3| Gene Symbol: Lysmd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030257.1 | NP_084533.1 | 3 | 878 | ||
| GenBank mRNA | AK029514.1 | - | 3 | 797 | ||
| BC003322.1 | - | 3 | 878 | 3156298 | ||