siRNA Details
| Gene Symbol: Slc19a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030556.2 | NP_085033.2 | 4 | 1066 | ||
| GenBank mRNA | AF271634.1 | - | 4 | 1066 | ||
| AK098112.1 | - | 4 | 1057 | |||
| BC109154.1 | - | 4 | 1049 | 40047017 | ||
| BC109155.1 | - | 3 | 919 | 40047019 | ||

Slc19a3
Genomic Map