siRNA Details
| Gene Symbol: Slc19a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030556.2 | NP_085033.2 | 6 | 1451 | ||
| GenBank mRNA | AF271634.1 | - | 6 | 1451 | ||
| AK079566.1 | - | 3 | 370 | |||
| AK098112.1 | - | 6 | 1442 | |||
| BC109154.1 | - | 6 | 1434 | 40047017 | ||
| BC109155.1 | - | 5 | 1304 | 40047019 | ||

Slc19a3
Genomic Map