siRNA Details
| Gene Symbol: BC004004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030561.3 | NP_085038.3 | 9 | 1302 | ||
| GenBank mRNA | AK030596.1 | - | 7 | 872 | ||
| AK151327.1 | - | 2 | 111 | |||
| AK211240.1 | - | 2 | 114 | |||
| BC058575.1 | - | 9 | 1111 | 6488611 | ||

BC004004
Genomic Map