siRNA Details
| Gene Symbol: Nfkbiz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159394.1 | NP_001152866.1 | 5 | 730 | ||
| NM_001159395.1 | NP_001152867.1 | 6 | 612 | |||
| NM_030612.3 | NP_085115.1 | 7 | 960 | |||
| GenBank mRNA | AB020974.1 | - | 7 | 1172 | ||
| AB026551.2 | - | 5 | 730 | |||
| AB047549.2 | - | 6 | 625 | |||
| AB196497.1 | - | 1 | 65 | |||
| AK082908.1 | - | 5 | 729 | |||
| AK136509.1 | - | 2 | 199 | |||
| AK149946.1 | - | 6 | 608 | |||
| AK150519.1 | - | 5 | 729 | |||
| AK152016.1 | - | 5 | 731 | |||
| AK156826.1 | - | 5 | 731 | |||
| AK159093.1 | - | 5 | 731 | |||
| AK159171.1 | - | 5 | 731 | |||
| AK159459.1 | - | 5 | 731 | |||
| AK159645.1 | - | 5 | 731 | |||
| AK159917.1 | - | 5 | 731 | |||
| AK160065.1 | - | 5 | 729 | |||
| BC058188.1 | - | 5 | 715 | 5341736 | ||

Nfkbiz
Genomic Map