siRNA Details
| Gene Symbol: Ghdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031871.1 | NP_114077.1 | 9 | 1505 | ||
| GenBank mRNA | AF316996.1 | - | 9 | 1505 | ||
| AK143922.1 | - | 9 | 1474 | |||
| AK161571.1 | - | 6 | 653 | |||
| BC005639.1 | - | 9 | 1486 | 3256858 | ||

Ghdc
Genomic Map