siRNA Details
| Gene Symbol: Ntng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163348.1 | NP_001156820.1 | 6, 7 | 2307 | ||
| NM_030699.2 | NP_109624.1 | 7, 8 | 2475 | |||
| GenBank mRNA | AB038666.1 | - | 5, 6 | 1212 | ||
| AB038667.1 | - | 7, 8 | 2252 | |||
| AF475069.1 | - | 6, 7 | 1380 | |||
| AF475070.1 | - | 5, 6 | 1212 | |||

Ntng1
Genomic Map