siRNA Details
| Gene Symbol: Zfhx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030708.2 | NP_109633.2 | 10 | 9523 | ||
| GenBank mRNA | AB024499.1 | - | 11 | 9054 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfhx4| Gene Symbol: Zfhx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030708.2 | NP_109633.2 | 10 | 9523 | ||
| GenBank mRNA | AB024499.1 | - | 11 | 9054 | ||