siRNA Details
| Gene Symbol: Abo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290444.1 | NP_001277373.1 | 5 | 763 | ||
| NM_030718.5 | NP_109643.3 | 6 | 898 | |||
| GenBank mRNA | AB041039.1 | - | 5 | 786 | ||
| AK033786.1 | - | 6 | 819 | |||
| AK035261.1 | - | 6 | 883 | |||
| BC116759.1 | - | 6 | 847 | 40126078 | ||
| BC116761.1 | - | 6 | 847 | 40126080 | ||
| DV046627.1 | - | 5 | 782 | |||

Abo
Genomic Map