siRNA Details
| Gene Symbol: Gatsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030719.3 | NP_109644.2 | 7, 8 | 1112 | ||
| GenBank mRNA | AJ296173.1 | - | 7, 8 | 1152 | ||
| AK039122.1 | - | 7, 8 | 1138 | |||
| AK052217.1 | - | 7, 8 | 1097 | |||
| BC094613.1 | - | 7, 8 | 1112 | 6856325 | ||

Gatsl2
Genomic Map