siRNA Details
| Gene Symbol: Slc26a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289787.1 | NP_001276716.1 | 15 | 1812 | ||
| NM_001289788.1 | NP_001276717.1 | 15 | 1760 | |||
| NM_030727.5 | NP_109652.3 | 17 | 1998 | |||
| GenBank mRNA | AF529192.1 | - | 17 | 1998 | ||
| AY024359.2 | - | 15 | 1764 | |||
| BC108986.1 | - | 15 | 1752 | 40047724 | ||
| BC108987.1 | - | 15 | 1804 | 40047725 | ||

Slc26a5
Genomic Map