siRNA Details
| Gene Symbol: Trim23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030731.3 | NP_109656.1 | 8, 9 | 3312 | ||
| GenBank mRNA | AK029533.1 | - | 9, 10 | 1461 | ||
| AK030843.1 | - | 9, 10 | 1460 | |||
| AK030862.1 | - | 8, 9 | 3313 | |||
| AK036210.1 | - | 9, 10 | 1461 | |||
| AK039280.1 | - | 9, 10 | 1472 | |||
| AK042070.1 | - | 9, 10 | 1463 | |||
| AK088922.1 | - | 8, 9 | 1277 | |||
| BC056390.1 | - | 9, 10 | 1453 | 6854944 | ||
| BC059017.1 | - | 9, 10 | 1453 | 6848690 | ||

Trim23
Genomic Map