siRNA Details
| Gene Symbol: Tbl1xr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030732.3 | NP_109657.2 | 4 | 504 | ||
| GenBank mRNA | AF268195.1 | - | 4 | 285 | ||
| AK029595.1 | - | 4 | 303 | |||
| AK033347.1 | - | 4 | 374 | |||
| AK036064.1 | - | 5 | 615 | |||
| AK136255.1 | - | 4 | 326 | |||

Tbl1xr1
Genomic Map