siRNA Details
| Gene Symbol: Rnf114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030743.5 | NP_109668.2 | 7 | 1449 | ||
| GenBank mRNA | AF282919.2 | - | 5 | 593 | ||
| AF502145.1 | - | 5 | 596 | |||
| AK079751.1 | - | 5 | 605 | |||
| AK088441.1 | - | 5 | 603 | |||
| AK139030.1 | - | 5 | 583 | |||
| AK148290.1 | - | 5 | 610 | |||
| AK148392.1 | - | 5 | 598 | |||
| AK150086.1 | - | 5 | 590 | |||
| AK150263.1 | - | 5 | 590 | |||
| AK150700.1 | - | 4 | 326 | |||
| AK152687.1 | - | 5 | 590 | |||
| AK153452.1 | - | 5 | 580 | |||
| AK159818.1 | - | 5 | 580 | |||
| AK163062.1 | - | 4 | 436 | |||
| BC023125.1 | - | 5 | 608 | |||
| BC043446.1 | - | 5 | 575 | |||
| BC054416.1 | - | 5 | 585 | 3672897 | ||
| BC085146.1 | - | 7 | 1449 | 6812845 | ||

Rnf114
Genomic Map