siRNA Details
| Gene Symbol: Sil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030749.2 | NP_109674.2 | 9 | 1123 | ||
| GenBank mRNA | AJ297884.1 | - | 9 | 1064 | ||
| AK170415.1 | - | 9 | 1076 | |||
| AK206841.1 | - | 1 | 15 | |||
| AK217356.1 | - | 2 | 224 | |||
| BC016119.1 | - | 9 | 1100 | 4489217 | ||
| BC016466.1 | - | 9 | 1086 | 3582798 | ||
| CO428760.1 | - | 6 | 666 | |||

Sil1
Genomic Map