siRNA Details
| Gene Symbol: Sgpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030750.3 | NP_109675.1 | 3 | 1208 | ||
| GenBank mRNA | AF247177.1 | - | 3 | 1158 | ||
| AF415215.1 | - | 3 | 1042 | |||
| AK149860.1 | - | 3 | 1209 | |||
| AK154116.1 | - | 4 | 850 | |||
| BC026814.1 | - | 3 | 1144 | |||
| BC037592.1 | - | 3 | 1161 | 5004335 | ||

Sgpp1
Genomic Map