siRNA Details
| Gene Symbol: Zbtb22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020625.3 | NP_065650.1 | 2 | 1667 | ||
| GenBank mRNA | AK003583.1 | - | 1 | 21 | ||
| AK155199.1 | - | 2 | 1651 | |||
| AK157161.1 | - | 2 | 1659 | |||
| AK159789.1 | - | 2 | 1658 | |||
| BC026964.1 | - | 2 | 1645 | 5043642 | ||
| BC052154.1 | - | 2 | 1614 | 6329405 | ||
| KJ650209.1 | - | 1 | 1503 | |||

Zbtb22
Genomic Map