siRNA Details
Gene Symbol: Zbtb22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020625.3 | NP_065650.1 | 2 | 1067 | ||
GenBank mRNA | AK155199.1 | - | 2 | 1051 | ||
AK157161.1 | - | 2 | 1059 | |||
AK159082.1 | - | 2 | 1057 | |||
AK159789.1 | - | 2 | 1058 | |||
BC026964.1 | - | 2 | 1045 | 5043642 | ||
BC052154.1 | - | 2 | 1014 | 6329405 | ||
KJ650209.1 | - | 1 | 903 |
