siRNA Details
Gene Symbol: Zbtb22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020625.3 | NP_065650.1 | 2 | 442 | ||
GenBank mRNA | AK155199.1 | - | 2 | 426 | ||
AK157161.1 | - | 2 | 433 | |||
AK159082.1 | - | 2 | 432 | |||
AK159789.1 | - | 2 | 433 | |||
BC026964.1 | - | 2 | 420 | 5043642 | ||
BC052154.1 | - | 2 | 389 | 6329405 | ||
KJ650209.1 | - | 1 | 278 |
