siRNA Details
| Gene Symbol: C1qtnf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204134.1 | NP_001191063.1 | 5 | 839 | ||
| NM_030888.4 | NP_112150.1 | 5 | 645 | |||
| GenBank mRNA | AF246265.1 | - | 5 | 622 | ||
| AK029352.1 | - | 5 | 622 | |||
| AK075867.1 | - | 5 | 622 | |||
| AK081704.1 | - | 5 | 622 | |||
| AK128898.1 | - | 5 | 645 | |||
| BC019636.1 | - | 5 | 848 | |||
| BC055047.1 | - | 5 | 607 | 6486507 | ||
| BC055859.1 | - | 5 | 586 | 4238617 | ||
| DQ002396.1 | - | 5 | 523 | |||

C1qtnf3
Genomic Map