siRNA Details
| Gene Symbol: C1qtnf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204134.1 | NP_001191063.1 | 3 | 542 | ||
| NM_030888.4 | NP_112150.1 | 3 | 348 | |||
| GenBank mRNA | AF246265.1 | - | 3 | 325 | ||
| AK029352.1 | - | 3 | 325 | |||
| AK075867.1 | - | 3 | 325 | |||
| AK081704.1 | - | 3 | 325 | |||
| AK128898.1 | - | 3 | 348 | |||
| BC019636.1 | - | 3 | 551 | |||
| BC055047.1 | - | 3 | 310 | 6486507 | ||
| BC055859.1 | - | 3 | 289 | 4238617 | ||
| DQ002396.1 | - | 3 | 226 | |||

C1qtnf3
Genomic Map