siRNA Details
| Gene Symbol: Ift122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167763.1 | NP_001161235.1 | 17 | 2086 | ||
| NM_031177.4 | NP_112454.2 | 17 | 2086 | |||
| GenBank mRNA | AK031308.1 | - | 17 | 1974 | ||
| AK044456.1 | - | 17 | 2086 | |||
| AK077104.1 | - | 4 | 549 | |||
| AK131115.1 | - | 17 | 2013 | |||
| AK167330.1 | - | 17 | 2041 | |||
| AK167412.1 | - | 17 | 2041 | |||
| BC038826.1 | - | 2 | 369 | |||
| BC060621.1 | - | 17 | 2173 | |||
| BC066083.1 | - | 17 | 2005 | 30355922 | ||

Ift122
Genomic Map